2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-((Ethoxythioxomethyl)thio)-2-methylpropanoic acid

A277456-1G A277456-250MG
76977-308EA 39.26 USD
76977-308 76977-310
2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid 97%
2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃S₂
MW: 208.29 g/mol
MDL Number: MFCD19689494
CAS Number: 133944-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR