5-(2-Fluorophenyl)-1,3,4-oxadiazol-2(3H)-one 98%

Supplier: Ambeed
Warning

A114136-1G A114136-250MG
76971-072EA 136.86 USD
76971-072 76971-074
5-(2-Fluorophenyl)-1,3,4-oxadiazol-2(3H)-one 98%
5-(2-Fluorophenyl)-1,3,4-oxadiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MW: 180.14 g/mol
MDL Number: MFCD11046702
CAS Number: 1044766-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR