2-Deoxy-2-fluoro-alpha-D-arabinofuranosyl bromide 3,5-dibenzoate 98%

Supplier: Ambeed
Warning

A261258-100MG A261258-250MG A261258-5G
76976-964EA 33.95 USD
76976-964 76976-966 77568-508
2-Deoxy-2-fluoro-alpha-D-arabinofuranosyl bromide 3,5-dibenzoate 98%
2-Deoxy-2-fluoro-alpha-D-arabinofuranosyl bromide 3,5-dibenzoate

Precaución: For research use only.

Formula: C₁₉H₁₆BrFO₅
MW: 423.22 g/mol
MDL Number: MFCD15144963
CAS Number: 97614-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR