4-Bromo-2-fluoro-6-methoxybenzonitrile 95%

Supplier: Ambeed
Warning

A329231-250MG A329231-1G A329231-100MG
76978-204EA 66 USD
76978-204 77593-946 77881-920
4-Bromo-2-fluoro-6-methoxybenzonitrile 95%
4-Bromo-2-fluoro-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MDL Number: MFCD16619400
CAS Number: 457051-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR