(2R,6R)-2,6-Dimethylpiperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A255625-250MG A255625-5G
76976-800EA 18 USD
76976-800 76976-802
(2R,6R)-2,6-Dimethylpiperazine dihydrochloride 97%
(2R,6R)-2,6-Dimethylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD13193063
CAS Number: 162240-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR