4-Chloro-2-iodo-1-nitrobenzene 97%

Supplier: Ambeed
Warning

A179084-10G A179084-1G A179084-250MG A179084-25G
76974-370EA 38 USD
76974-370 76974-372 76974-374 76974-376
4-Chloro-2-iodo-1-nitrobenzene 97%
4-Chloro-2-iodo-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MDL Number: MFCD11007732
CAS Number: 160938-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR