4-Fluoro-2-iodo-1-nitrobenzene 96%

Supplier: Ambeed
Warning

A264870-100G A264870-10G A264870-1G A264870-250MG A264870-25G A264870-5G
76977-052EA 2153.63 USD
76977-052 76977-054 76977-056 76977-058 76977-060 76977-062
4-Fluoro-2-iodo-1-nitrobenzene 96%
4-Fluoro-2-iodo-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 267.00 g/mol
MDL Number: MFCD16658618
CAS Number: 41860-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR