4-Chloro-5-fluoro-2-nitrophenol 95%

Supplier: Ambeed
Danger

A309662-100MG A309662-1G A309662-250MG
76977-878EA 14 USD
76977-878 76977-880 76977-882
4-Chloro-5-fluoro-2-nitrophenol 95%
4-Chloro-5-fluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₃
MDL Number: MFCD11840294
CAS Number: 345-25-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR