1-(4-Chlorophenyl)cyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A440749-100G A440749-1G A440749-25G A440749-5G
76979-376EA 530 USD
76979-376 76979-378 76979-380 76979-382
1-(4-Chlorophenyl)cyclobutanecarboxylic acid 98%
1-(4-Chlorophenyl)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₂
MDL Number: MFCD00001324
CAS Number: 50921-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR