5-Bromo-7-iodo-1H-indazole 95%

Supplier: Ambeed
Warning

A179902-250MG A179902-100MG
76974-414EA 88 USD
76974-414 77881-364
5-Bromo-7-iodo-1H-indazole 95%

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MDL Number: MFCD13177018
CAS Number: 953410-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR