N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A187704-100MG
A187704-10MG
A187704-50MG
A187704-5MG
76974-712EA
90
USD
76974-712
76974-714
76974-716
76974-718
76974-720
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine hydrochloride 98%
N-(3-Bromophenyl)-6,7-dimethoxyquinazolin-4-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₆H₁₄BrN₃O₂·HCl |
MDL Number:
MFCD01074805 CAS Number: 153436-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...