(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene

A792836-250MG A792836-5G
76981-628EA 25.47 USD
76981-628 76981-630
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%, ee 99%
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine

Precaución: for Research Use Only

Formula: C₃₂H₄₀FeP₂
MW: 542.4500 g/mol
MDL Number: MFCD01074442
CAS Number: 155830-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR