5-Methylthiazole-4-carboxylic acid ethyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-methylthiazole-4-carboxylate
A317646-1G
A317646-250MG
A317646-5G
76978-012EA
71.09
USD
76978-012
76978-014
76978-016
5-Methylthiazole-4-carboxylic acid ethyl ester 97%
5-Methylthiazole-4-carboxylic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₇H₉NO₂S MW: 171.22 g/mol |
MDL Number:
MFCD18427704 CAS Number: 61323-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...