tert-Butyl (4-oxo-1,2,3,4-tetrahydronaphthalen-1-yl)carbamate 95%

Supplier: Ambeed
Warning

A186385-100MG A186385-1G A186385-250MG
76974-656EA 28 USD
76974-656 76974-658 76974-660
tert-Butyl (4-oxo-1,2,3,4-tetrahydronaphthalen-1-yl)carbamate 95%
tert-Butyl (4-oxo-1,2,3,4-tetrahydronaphthalen-1-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃
MDL Number: MFCD19689457
CAS Number: 1313021-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR