4-Methylpiperidin-4-ol hydrochloride 98%

Supplier: Ambeed
Warning

A144834-1G A144834-5G
76972-780EA 21.22 USD
76972-780 76972-782
4-Methylpiperidin-4-ol hydrochloride 98%
4-Methylpiperidin-4-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD11044137
CAS Number: 586375-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR