2-Amino-2-thiazoline 97%

Supplier: Ambeed
Warning

A635887-100G A635887-1G A635887-25G A635887-5G
76980-706EA 318 USD
76980-706 76980-708 76980-710 76980-712
2-Amino-2-thiazoline 97%
2-Amino-2-thiazoline

Precaución: for Research Use Only

Formula: C₃H₆N₂S
MDL Number: MFCD00005313
CAS Number: 1779-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR