Emricasan 98%

Supplier: Ambeed
Danger

Synonyms: (S)-3-((S)-2-(2-((2-(tert-Butyl)phenyl)amino)-2-oxoacetamido)propanamido)-4-oxo-5-(2,3,5,6-tetrafluorophenoxy)pentanoic acid

A622861-100MG A622861-250MG A622861-50MG
76980-636EA 431 USD
76980-636 77613-934 77613-936
Emricasan 98%
Emricasan

Precaución: for Research Use Only

Formula: C₂₆H₂₇F₄N₃O₇
MW: 569.50 g/mol
MDL Number: MFCD11840420
CAS Number: 254750-02-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR