4-(Cyanomethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A515096-10G A515096-1G A515096-250MG A515096-5G
76979-970EA 375.56 USD
76979-970 76979-972 76979-974 76979-976
4-(Cyanomethyl)benzoic acid 97%
4-(Cyanomethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD00020382
CAS Number: 50685-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR