2-(Methylcarboxy)pyridine-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl pyridine-5-boronic acid pinacol ester-2-carboxylate, Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

A564950-100MG A564950-1G A564950-250MG A564950-25G A564950-5G
76980-288EA 6 USD
76980-288 76980-290 76980-292 76980-294 76980-296
2-(Methylcarboxy)pyridine-5-boronic acid pinacol ester 97%
2-(Methylcarboxy)pyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MW: 263.1 g/mol
MDL Number: MFCD08063051
CAS Number: 957065-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR