Pyridazine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridazinecarboxylic acid

A160094-10G A160094-1G A160094-250MG A160094-25G A160094-5G
76973-518EA 303.42 USD
76973-518 76973-520 76973-522 76973-524 76973-526
Pyridazine-4-carboxylic acid 97%
Pyridazine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
MDL Number: MFCD00011576
CAS Number: 50681-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR