Methyl-6-chloro-5-methoxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-5-methoxynicotinate

A206040-100MG A206040-10G A206040-1G A206040-250MG A206040-5G
76975-338EA 21 USD
76975-338 76975-340 76975-342 76975-344 77671-820
Methyl-6-chloro-5-methoxynicotinate 98%
Methyl-6-chloro-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MDL Number: MFCD16659999
CAS Number: 915107-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR