5-Bromo-2-fluoro-3-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A814097-1G A814097-250MG A814097-5G A814097-25G
76981-700EA 40 USD
76981-700 76981-702 76981-704 77882-736
5-Bromo-2-fluoro-3-methylbenzaldehyde 98%
5-Bromo-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD11110261
CAS Number: 903875-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR