6-Bromo-3-hydroxyquinoline 98%

Supplier: Ambeed
Warning

A292550-100MG A292550-1G A292550-250MG
76977-610EA 25 USD
76977-610 76977-612 76977-614
6-Bromo-3-hydroxyquinoline 98%
6-Bromo-3-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD09260430
CAS Number: 552330-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR