(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(piperidin-1-yl)methanone 98+%

Supplier: Ambeed
Warning

Synonyms: CX546

A118774-10MG A118774-25MG A118774-50MG A118774-1MG
76971-368EA 42.44 USD
76971-368 76971-370 76971-372 77610-978
(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(piperidin-1-yl)methanone 98+%
(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD01860868
CAS Number: 215923-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR