5-Fluoro-2-methylpyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-methylpyrimidin-4-ol

A159728-250MG A159728-5G
76973-488EA 40.32 USD
76973-488 76973-490
5-Fluoro-2-methylpyrimidin-4(3H)-one 95%
5-Fluoro-2-methylpyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₅H₅FN₂O
MW: 128.11 g/mol
MDL Number: MFCD16988103
CAS Number: 1480-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR