4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(Dimethylamino)methyl]benzeneboronic acid pinacol ester, Dimethyl-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzyl]amine, N,N-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine

A953505-1G A953505-250MG A953505-5G
76982-010EA 55 USD
76982-010 76982-012 76982-014
4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester 98%
4-[(Dimethylamino)methyl]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₂
MW: 261.168 g/mol
MDL Number: MFCD04971190
CAS Number: 878197-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR