1,13-Tridecanedioic acid 95%

Supplier: Ambeed
Warning

Synonyms: Tridecanedioic acid, Undecane-1,11-dicarboxylic acid, 1,11-Undecanedicarboxylic acid

A170816-100G A170816-10G A170816-1G A170816-25G A170816-500G A170816-5G
76973-990EA 49.87 USD
76973-990 76973-992 76973-994 76973-996 76973-998 76974-000
1,13-Tridecanedioic acid 95%
1,13-Tridecanedioic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₄
MW: 244.33 g/mol
MDL Number: MFCD00002740
CAS Number: 505-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR