5,7-Dichloropyrido[3,4-b]pyrazine 97%

Supplier: Ambeed
Warning

A125181-1G A125181-250MG
76971-806EA 158.08 USD
76971-806 76971-808
5,7-Dichloropyrido[3,4-b]pyrazine 97%
5,7-Dichloropyrido[3,4-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MW: 200.03 g/mol
MDL Number: MFCD19707629
CAS Number: 1379338-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR