2-Bromo-1-isopropyl-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A180307-10G A180307-1G A180307-5G
76974-438EA 237 USD
76974-438 76974-440 76974-442
2-Bromo-1-isopropyl-4-nitrobenzene 97%
2-Bromo-1-isopropyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD12024309
CAS Number: 101980-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR