4-Chloro-2-hydroxybenzonitrile 95%
Supplier: Ambeed
|
Warning
|
A252451-250MG
A252451-1G
76976-710EA
13.8
USD
76976-710
77672-260
4-Chloro-2-hydroxybenzonitrile 95%
4-Chloro-2-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₄ClNO MW: 153.5700 g/mol |
MDL Number:
MFCD00234255 CAS Number: 30818-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...