1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A251422-10G A251422-1G A251422-250MG A251422-5G A251422-25G
76976-692EA 387.23 USD
76976-692 76976-694 76976-696 77612-424 77672-238
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₂
MW: 234.106 g/mol
MDL Number: MFCD16659007
CAS Number: 1151802-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR