Ponatinib 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-benzamide, 3-(Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide, AP24534

A485520-1MG A485520-1G A485520-5MG A485520-250MG A485520-10MG
76979-772EA 5.31 USD
76979-772 77594-510 77613-522 77673-268 77673-266
Ponatinib 98%
Ponatinib

Precaución: for Research Use Only

Formula: C₂₉H₂₇F₃N₆O
MW: 532.56 g/mol
MDL Number: MFCD17215203
CAS Number: 943319-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR