1-Phenylpiperidin-4-amine 97%

Supplier: Ambeed
Warning

A664180-100MG A664180-1G A664180-250MG A664180-5G
76980-872EA 40 USD
76980-872 76980-874 76980-876 76980-878
1-Phenylpiperidin-4-amine 97%
1-Phenylpiperidin-4-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MDL Number: MFCD00044398
CAS Number: 63921-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR