Boc-His(Z)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Boc-His(Cbz)-OH

A678736-100MG A678736-10G A678736-250MG A678736-25G A678736-5G
76980-988EA 6.37 USD
76980-988 76980-990 76980-992 76980-994 76980-996
Boc-His(Z)-OH 97%
Boc-His(Z)-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₃O₆
MW: 389.408 g/mol
MDL Number: MFCD00076942
CAS Number: 50305-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR