Bromotris(dimethylamino)phosphonium hexafluorophosphate 98%

Supplier: Ambeed
Danger

Synonyms: Bromotris(dimethylamino)phosphonium Hexafluorophosphate

A497772-1G A497772-5G A497772-25G
76979-844EA 10 USD
76979-844 76979-846 77572-580
Bromotris(dimethylamino)phosphonium hexafluorophosphate 98%
Bromotris(dimethylamino)phosphonium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₆H₁₈BrF₆N₃P₂
MW: 388.07 g/mol
MDL Number: MFCD00191864
CAS Number: 50296-37-2
UN: 3263
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR