5-Isopropylthiazol-2-amine 97%

Supplier: Ambeed
Warning

A284496-250MG A284496-5G A284496-1G
76977-486EA 55 USD
76977-486 76977-488 77881-775
5-Isopropylthiazol-2-amine 97%
5-Isopropylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂S
MDL Number: MFCD13185908
CAS Number: 101080-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR