3-Bromo-4-iodoaniline 97%

Supplier: Ambeed
Warning

A774777-1G A774777-250MG A774777-5G
76981-546EA 16 USD
76981-546 76981-548 76981-550
3-Bromo-4-iodoaniline 97%
3-Bromo-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MDL Number: MFCD09753717
CAS Number: 860435-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR