8-Bromoquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A325261-100MG A325261-1G A325261-250MG
76978-142EA 63.66 USD
76978-142 76978-144 76978-146
8-Bromoquinoline-3-carboxylic acid 98%
8-Bromoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.0641 g/mol
MDL Number: MFCD18375331
CAS Number: 347146-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR