2-Chloroquinoline-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A104040-1G A104040-5G
76970-310EA 38.2 USD
76970-310 76970-312
2-Chloroquinoline-5-carboxylic acid 98%
2-Chloroquinoline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MW: 207.6100 g/mol
MDL Number: MFCD11226761
CAS Number: 1092287-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR