(1R,2R)-N1,N2-Dibenzyl-1,2-diphenylethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A176194-100MG A176194-250MG
76974-260EA 21.22 USD
76974-260 76974-262
(1R,2R)-N1,N2-Dibenzyl-1,2-diphenylethane-1,2-diamine 97%
(1R,2R)-N1,N2-Dibenzyl-1,2-diphenylethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₈H₂₈N₂
MW: 392.54 g/mol
CAS Number: 221226-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR