(2-Fluoro-6-methoxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A148958-1G A148958-250MG
76973-022EA 24.41 USD
76973-022 76973-024
(2-Fluoro-6-methoxyphenyl)acetic acid 95%
(2-Fluoro-6-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.16 g/mol
MDL Number: MFCD00671770
CAS Number: 500912-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR