(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A439331-100G
A439331-250MG
A439331-5G
A439331-25G
A439331-1G
76979-348EA
388
USD
76979-348
76979-350
76979-352
77568-874
77613-346
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥97%, ee 99%
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Precaución: For research use only.
Formula:
C₂₄H₂₂O₄ |
MDL Number:
MFCD03788935 CAS Number: 142128-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...