(S)-(+)-(8,9,10,11,12,13,14,15-Octahydro-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-N,N-Dimethyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine
A264873-100MG
A264873-250MG
76977-064EA
61.54
USD
76977-064
76977-066
(S)-(+)-(8,9,10,11,12,13,14,15-Octahydro-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98%, ee 99%
(S)-(+)-(8,9,10,11,12,13,14,15-Octahydro-3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine
Precaución: for Research Use Only
Formula:
C₂₂H₂₆NO₂P MW: 367.4211 g/mol |
MDL Number:
MFCD04117692 CAS Number: 389130-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...