(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine 95%

Supplier: Ambeed
Warning

A309311-250MG A309311-5G A309311-10G A309311-1G A309311-25G
76977-866EA 6 USD
76977-866 76977-868 77881-860 77881-861 77881-862
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine 95%
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine

Precaución: for Research Use Only

Formula: C₂₂H₁₈NO₂P
MDL Number: MFCD03426988
CAS Number: 185449-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR