9-(1-Naphthyl)anthracene 99%

Supplier: Ambeed
Warning

Synonyms: 9-(Naphthalen-1-yl)anthracene

A948747-1G A948747-5G
76982-006EA 18.04 USD
76982-006 76982-008
9-(1-Naphthyl)anthracene 99%
9-(1-Naphthyl)anthracene

Precaución: for Research Use Only

Formula: C₂₄H₁₆
MW: 304.39 g/mol
MDL Number: MFCD19689379
CAS Number: 7424-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR