2-Morpholino-4-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine

A773374-1G A773374-250MG A773374-5G
76981-522EA 166 USD
76981-522 76981-524 76981-526
2-Morpholino-4-pyridineboronic acid pinacol ester 98%
2-Morpholino-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₃
MDL Number: MFCD11054039
CAS Number: 888721-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR