Methyl (S)-(+)-2-pyrrolidone-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, H-Pyr-OMe

A234185-100G A234185-10G A234185-500G A234185-5G A234185-25G A234185-1000G
76976-252EA 36.08 USD
76976-252 76976-254 76976-256 76976-258 77089-664 77672-086
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate 95%
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.14 g/mol
MDL Number: MFCD00080931
CAS Number: 4931-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR