2-(2-Nitroanilino)ethanol 98%

Supplier: Ambeed
Warning

A426829-100G A426829-10G
76979-284EA 33 USD
76979-284 76979-286
2-(2-Nitroanilino)ethanol 98%
2-(2-Nitroanilino)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD00239475
CAS Number: 4926-55-0
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR