1,4,7-Triazacyclononane trihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1,4,7-Triazonane trihydrochloride

A145596-250MG A145596-5G A145596-1G A145596-25G A145596-100G
76972-824EA 9.55 USD
76972-824 76972-826 77592-722 77592-724 77611-344
1,4,7-Triazacyclononane trihydrochloride 97%
1,4,7-Triazacyclononane trihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₈Cl₃N₃
MW: 238.5900 g/mol
MDL Number: MFCD00074887
CAS Number: 58966-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR