1,2,3,4-Tetrahydro-1-methylquinoline 95%

Supplier: Ambeed
Warning

A406811-1G A406811-250MG A406811-5G
76979-116EA 35.01 USD
76979-116 76979-118 76979-120
1,2,3,4-Tetrahydro-1-methylquinoline 95%
1,2,3,4-Tetrahydro-1-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MDL Number: MFCD00130001
CAS Number: 491-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR